Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

904953

Sigma-Aldrich

[Ru(dtbbpy)3](PF6)2

≥95%

Synonym(s):

Tris(4,4′-di-tert-butyl-2,2′-bipyridine)ruthenium bis(hexafluorophosphate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H72F12N6P2Ru
CAS Number:
Molecular Weight:
1196.19
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

reaction suitability

core: ruthenium
reagent type: catalyst
reaction type: Photocatalysis

mp

>300 °C

InChI key

XBDHEZBIYYTEHN-UHFFFAOYSA-N

Application

This ruthenium photocatalyst facilitates a variety of transformations using visible light.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service