Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

902853

Sigma-Aldrich

Metoprolol acid

≥95%

Synonym(s):

4-(2-Hydroxy-3-isopropylaminopropoxy)phenylacetic acid, 4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetic acid, Atenolol acid, Atenolol impurity G (EP), H 117/04, SL 77-010

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21NO4
CAS Number:
Molecular Weight:
267.32
MDL number:
UNSPSC Code:
12352112
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

storage temp.

−20°C

InChI

1S/C14H21NO4/c1-10(2)15-8-12(16)9-19-13-5-3-11(4-6-13)7-14(17)18/h3-6,10,12,15-16H,7-9H2,1-2H3,(H,17,18)

InChI key

PUQIRTNPJRFRCZ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service