Skip to Content
MilliporeSigma
All Photos(1)

Documents

902527

Sigma-Aldrich

Amino-PEG5-t-butyl ester

≥95%

Synonym(s):

tert-Butyl-1-amino-3,6,9,12,15-pentaoxaoctadecan-18-oate, H2N-PEG5-CH2CH2COOtBu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H35NO7
CAS Number:
Molecular Weight:
365.46
MDL number:
UNSPSC Code:
51171641

Assay

≥95%

form

liquid

reaction suitability

reagent type: cross-linking reagent

refractive index

n/D 1.4534

density

1.4690 g/mL

functional group

amine
ester

storage temp.

−20°C

SMILES string

NCCOCCOCCOCCOCCOCCC(OC(C)(C)C)=O

Application

This heterobifunctional, PEGylated crosslinker features an amino group at one end and t-butyl-protected carboxyl group at the other, which can be deprotected with acidic conditions. The hydrophillic PEG linker facilitates solubility in biological applications. Amino-PEG5-t-butyl ester can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or proteolysis-targeting chimeras (PROTAC® molecules) for targeted protein degradation.

Technology Spotlight: Degrader Building Blocks for Targeted Protein Degradation

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service