Skip to Content
MilliporeSigma
All Photos(2)

Documents

901931

Sigma-Aldrich

NORIT® A SUPRA EUR USP

meets analytical specifications of USP

Synonym(s):

Activated carbon, Activated charcoal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C
CAS Number:
UNSPSC Code:
23201100
NACRES:
NA.23

description

Apparent Density, tamped: 420 kg/m3
Ash content: 2 mass %
Filtration time: 40 min
Iodine number: 1550
Methylene blue adsorption: 41 g/100g
Molasses number: 430 (EUR)

form

powder

quality

meets analytical specifications of USP

surface area

1700 m2/g , Total

particle size

100 μm (D90)
20 μm (D50)
4 μm (D10)

pH

neutral

bulk density

350‑450  kg/m3

anion traces

chloride (Cl-): 0.1 mass% (acid extr.)

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Application

  • Activated carbons are internally porous microcrystalline, non-graphitic forms of carbon. The activated carbons possess large surface area (~1000 m2/g) and pore volumes which make them useful for applications in energy storage devices, catalysis, and for removal of impurities from gases and liquids.
  • NORIT® A SUPRA EUR USP exhibits high adsorption capacity and is well suited for use as an adsorbent for the purification of pharmaceutical products. It also finds application as as antidote in case of poisoning. NORIT® A SUPRA EUR USP is a medicinal, steam activated carbon of 100% natural origin.

Legal Information

Product of Cabot Corporation.
NORIT® is a registered trademark of Cabot Corporation.
Norit is a registered trademark of Norit N.V.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Water and Wastewater Treatment: Historical Perspective of Activated Carbon Adsorption and its Integration with Biological Processes.
Aktas O, et al.
Wiley-VCH (2011)
Energy storage applications of activated carbons: supercapacitors and hydrogen storage.
Sevilla M and Mokaya R
Energy & Environmental Science, 7, 1250-1280 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service