Skip to Content
MilliporeSigma
All Photos(3)

Documents

901019

Sigma-Aldrich

Carbon nanotube, multi-walled

50-90 nm diameter, >95% carbon basis

Synonym(s):

MWCNT, MWNT, Multiwall carbon nanotube

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

description

D/G ratio: 0.1 (by Raman spectroscopy)
Real density: 2.1 g/cm^3 (by Pycnometer)

Quality Level

Assay

>95% carbon basis

aspect ratio

>100 , by FE-SEM

surface area

28 m2/g , by N<SUB>2</SUB> adsorption

diameter

50-90 nm
65 nm (by FE-SEM)

mp

3652-3697 °C (lit.)

density

~2.1 g/mL at 25 °C (lit.)

bulk density

0.007 g/cm3 (by HCC method)

Looking for similar products? Visit Product Comparison Guide

General description

Carbon nanotubes are hollow cylinders of carbon atoms. They are rolled-upgraphene sheets with carbon atoms covalently bonded to each other through sp2hybridization. Owing to their excellent mechanical and electrical properties,they are widely used in the field of flexible and wearable electronicdevices.

Application

Multi-walled carbon nanotubes (MWCNTs) functionalized with a polymer blend can be used as a hybrid catalyst for the degradation of crystal violet under visible light.

Due to their high magnetic sensitivity, MWCNT/manganese–zinc ferrite nanocomposites can be used in integrated nanoscale electronic systems.

They can also be used tofabricate sensors for strain-sensing applications because of their highmechanical flexibility and stability.

Features and Benefits

  • Excellent conductivity
  • Light weight
  • Chemical stability
  • Large surface area
  • High mechanical strength

Preparation Note

Chemical Vapor Deposition (CVD) Method

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service