Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

900913

Sigma-Aldrich

Tetrazine-PEG5-NHS ester

≥95%

Synonym(s):

Tz-PEG5-NHS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H36N6O10
CAS Number:
Molecular Weight:
604.61
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: click chemistry
reagent type: linker

functional group

NHS ester

storage temp.

−20°C

SMILES string

O=C(NCC1=CC=C(C2=NN=CN=N2)C=C1)CCOCCOCCOCCOCCOCCC(ON3C(CCC3=O)=O)=O

InChI

1S/C27H36N6O10/c34-23(28-19-21-1-3-22(4-2-21)27-31-29-20-30-32-27)7-9-38-11-13-40-15-17-42-18-16-41-14-12-39-10-8-26(37)43-33-24(35)5-6-25(33)36/h1-4,20H,5-19H2,(H,28,34)

InChI key

DUPHIQYIPUTDJS-UHFFFAOYSA-N

General description

Tetrazine-PEG5-NHS ester has an N-hydroxysuccinimide (NHS) ester that can be reacted with primary amines and tetrazine, which is reactive to trans-cyclooctenes. Hydrogen substituted tetrazines demonstrate exceptionally fast kinetics, generally at least 10-fold faster compared to methyl substituted tetrazines. The aqueous solubility of this reagent is substantially enhanced by a hydrophilic polyethylene glycol (PEG) spacer arm.

Application

Tetrazine-PEG5-NHS ester (Tz-PEG5-NHS) can be used in click chemistry applications[1][2] such as the preparation of chitosan-polyacrylamide microspheres utilizing selective tetrazine-trans-cyclooctene (Tz-TCO) ligation[1][3] and the construction of copper-64 radiolabeled antibody by the click reaction.[2]

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Porosity-Tuned Chitosan?Polyacrylamide Hydrogel Microspheres for Improved Protein Conjugation.
Jung S, et al.
Biomacromolecules, 17(7), 2427-2436 (2016)
Shape-Encoded Chitosan?Polyacrylamide Hybrid Hydrogel Microparticles with Controlled Macroporous Structures via Replica Molding for Programmable Biomacromolecular Conjugation.
Kang E, et al.
Langmuir, 32(21), 5394-5402 (2016)
Amit Kumar et al.
Bioconjugate chemistry, 26(4), 782-789 (2015-03-12)
We report a click-chemistry based modular strategy for antibody labeling with (64)Cu (t1/2 = 12.7 h; β(+) 0.656 MeV, 17.4%; β(-) 0.573 MeV, 39%; EC 43%) under ambient condition utilizing a cross-bridged tetraazamacrocyclic (CB-TE2A) analogue, which otherwise requires harsh conditions

Articles

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service