Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900748

Sigma-Aldrich

Perovskite quantum dots

oleic acid and oleylamine coated, fluorescence λem 450 nm, 10 mg/mL in toluene

Synonym(s):

Cadmium free QDs, Fluorescent nanocrystals, Perovskite nanocrystals, QDs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

Quality Level

form

liquid

concentration

10 mg/mL in toluene

fluorescence

λem 450 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

For Perovskite quantum dots:
Perovskite quantum dots (QDs) of common formula CsPbX3 (X = Cl, Br, I) possess high photoluminescence efficiency and narrow emission and emit in the visible spectral regime. Perovskite QDs are cadmium free and the aforementioned properties render them suitable for applications in light emitting diodes (LEDs), lasers, liquid crystal displays (LCDs) etc.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 1A - Skin Irrit. 2 - STOT RE 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

48.2 °F

Flash Point(C)

9 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

CsPbX3 Quantum Dots for Lighting and Displays: Room-Temperature Synthesis, Photoluminescence Superiorities, Underlying Origins and White Light-Emitting Diodes.
Li X, et al.
Advances in Functional Materials, 26(16), 2435-2445 (2016)
Loredana Protesescu et al.
Nano letters, 15(6), 3692-3696 (2015-01-31)
Metal halides perovskites, such as hybrid organic-inorganic CH3NH3PbI3, are newcomer optoelectronic materials that have attracted enormous attention as solution-deposited absorbing layers in solar cells with power conversion efficiencies reaching 20%. Herein we demonstrate a new avenue for halide perovskites by

Articles

In this article, the properties of some of the new non-cadmium based QDs along with different applications of QDs are summarized.

In this article, the properties of some of the new non-cadmium based QDs along with different applications of QDs are summarized.

In this article, the properties of some of the new non-cadmium based QDs along with different applications of QDs are summarized.

In this article, the properties of some of the new non-cadmium based QDs along with different applications of QDs are summarized.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service