Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

900608

Sigma-Aldrich

APhos Pd G4

95%

Synonym(s):

(Methanesulfonato-κO)[2′-(methylamino)-2-biphenylyl]palladium - 4-[bis(2-methyl-2-propanyl)phosphino]-N,N-dimethylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H43N2O3PPdS
CAS Number:
Molecular Weight:
649.13
UNSPSC Code:
12161600
NACRES:
NA.22

Quality Level

Assay

95%

form

powder or crystals

feature

generation 4

reaction suitability

core: palladium
reagent type: catalyst
reaction type: Cross Couplings

mp

204 °C

SMILES string

O=S(=O)([O-][Pd+2]1([C-]=2C=CC=CC2C=3C=CC=CC3[NH]1C)[P](C4=CC=C(C=C4)N(C)C)(C(C)(C)C)C(C)(C)C)C

InChI

1S/C16H28NP.C13H12N.CH4O3S.Pd/c1-15(2,3)18(16(4,5)6)14-11-9-13(10-12-14)17(7)8;1-14-13-10-6-5-9-12(13)11-7-3-2-4-8-11;1-5(2,3)4;/h9-12H,1-8H3;2-7,9-10,14H,1H3;1H3,(H,2,3,4);/q;-1;;+2/p-1

InChI key

MLEINNWTBGCQPS-UHFFFAOYSA-M

Application

APhos Pd G4 is a powerful ligand for classic cross-coupling reactions combined with the Buchwald Fourth Generation Palladacycle. Bench stable, soluble in most organic solvents.

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathaniel H Park et al.
Angewandte Chemie (International ed. in English), 54(28), 8259-8262 (2015-06-03)
In Pd-catalyzed C-N cross-coupling reactions, α-branched secondary amines are difficult coupling partners and the desired products are often produced in low yields. In order to provide a robust method for accessing N-aryl α-branched tertiary amines, new catalysts have been designed

Articles

G3 and G4 Buchwald palladium precatalysts are the newest air, moisture, and thermally stable crossing-coupling complexes used in bond formation for their versatility and high reactivity.

Related Content

The Buchwald group has developed a series of highly active and versatile palladium precatalysts and biarylphosphine ligands used in cross-coupling reactions for the formation of C-C, C–N, C–O, C–F, C–CF3, and C–S bonds. The ligands are electron-rich, and highly tunable to provide catalyst systems with a diverse scope, high stability and reactivity. Furthermore, the new series of precatalysts are air-, moisture and thermally-stable and display good solubility in common organic solvents. The use of precatalysts ensures the efficient generation of the active catalytic species and allows one to accurately adjust the ligand:palladium ratio. The ligands, precatalysts and methodology developed in the Buchwald group are user friendly and have rendered previously difficult cross couplings reactions, much easier to achieve.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service