Skip to Content
MilliporeSigma
All Photos(1)

Documents

900535

Sigma-Aldrich

Doped graphene

boron/nitrogen-doped, avg. no. of layers, 1 ‑ 5

Synonym(s):

B/N co-doped graphene, BNG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141908
NACRES:
NA.23

product name

Boron/Nitrogen co-doped graphene, avg. no. of layers, 1 ‑ 5

description

Oxygen: <7.5%

Quality Level

Assay

≥95%

form

powder

feature

avg. no. of layers 1 ‑ 5

surface area

>500 m2/g

cation traces

B: 2.0-4.0%
C: 85-95%
N: 2.0-4.0%

General description

  • Typical thickness: 1-5 layers.
  • Typical size : 0.5-5 μm.

Application

  • Electro catalyst.
  • Field-effect transistors.
  • Sensors.
  • Lithium ion batteries.
  • Supercapacitors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, Structure, and Properties of Boron- and Nitrogen-Doped Graphene.
Panchakarla LS, et al.
Advanced Materials, 21(46), 4726-4730 (2009)
Three-dimensional nitrogen and boron co-doped graphene for high-performance all-solid-state supercapacitors.
Zhong-Shuai Wu et al.
Advanced materials (Deerfield Beach, Fla.), 24(37), 5130-5135 (2012-07-19)
Boron-doped, nitrogen-doped, and codoped graphene on Cu(111): A DFT + vdW Study.
Ferrighi L, et al.
The Journal of Physical Chemistry C, 119(11), 6056-6064 (2015)

Articles

Advanced technologies for energy conversion and storage aim to improve performance and reduce environmental impact.

Advanced technologies for energy conversion and storage aim to improve performance and reduce environmental impact.

Advanced technologies for energy conversion and storage aim to improve performance and reduce environmental impact.

Advanced technologies for energy conversion and storage aim to improve performance and reduce environmental impact.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service