Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900397

Sigma-Aldrich

SnAP-ex 3-O-MOM P

95%

Synonym(s):

4-(Methoxymethoxy)-4-(tributylstannyl)butan-1-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H41NO2Sn
Molecular Weight:
398.21
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

paste

refractive index

n/D 1.4933

density

1.1407 g/mL

functional group

amine
ether

shipped in

wet ice

storage temp.

−20°C

Application

[SnAP reagents] provide a one-step route, in tandem with various aldehyde substrates, to saturated N-heterocycles. The synthesis of N-heterocycles through SnAP reagents requires mild reaction conditions, and aldehydes bearing aryl, heteroaryl, glyoxyl, aliphatic, and halogenated groups are well tolerated. This product was introduced in collaboration with the [Bode research group].

related product

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SnAP-eX reagents for the synthesis of exocyclic 3-amino-and 3-alkoxypyrrolidines and piperidines from aldehydes.
Luescher M U and Bode J W
Organic Letters, 18(11), 2652-2655 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service