Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

85150

Sigma-Aldrich

Silver carbonate

purum p.a., ≥99.0% (AT)

Synonym(s):

Disilver carbonate, Silver(I) carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag2CO3
CAS Number:
Molecular Weight:
275.75
Beilstein:
6936654
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

grade

purum p.a.

Quality Level

Assay

≥99.0% (AT)

form

powder

reaction suitability

reagent type: catalyst
core: silver

mp

210 °C (dec.) (lit.)

density

6.08 g/mL at 25 °C (lit.)

anion traces

sulfate (SO42-): ≤500 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Ag]OC(=O)O[Ag]

InChI

1S/CH2O3.2Ag/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

KQTXIZHBFFWWFW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Silver carbonate is used in Koenigs-Knorr glycosylation.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gregory L Hamilton et al.
Journal of the American Chemical Society, 130(45), 14984-14986 (2008-10-22)
Reactions proceeding through cationic intermediates that lack a Lewis or Brønsted basic site present a challenge for traditional asymmetric catalysis based on chiral metals or organocatalysts. We present an enantioselective ring opening of tetrasubstituted meso-aziridinium ions with alcohol nucleophiles proceeding
Palladium-catalyzed decarboxylative C-H bond arylation of thiophenes.
Peng Hu et al.
Angewandte Chemie (International ed. in English), 51(1), 227-231 (2011-11-17)
Roman V Rozhkov et al.
The Journal of organic chemistry, 68(16), 6314-6320 (2003-08-05)
The palladium-catalyzed annulation of 1,3-dienes by o-iodoacetoxycoumarins provides an efficient method for the synthesis of biologically interesting dihydrofurocoumarins. The presence of the acetyl group on the phenolic oxygen and the use of silver carbonate as a base are crucial for
Satish Malik et al.
Carbohydrate research, 345(5), 559-564 (2010-02-05)
A simple, mild, and regioselective method has been developed for the selective benzylation and p-methoxybenzylation of carbohydrate derivatives in high yields using Ag(2)CO(3) as the promoter. Benzylation of base-labile substrates, for which other reported methods are of little use, has
K Y Jan et al.
Experimental cell research, 159(1), 55-62 (1985-07-01)
Opposite differential staining between sister chromatids was obtained by two silver-staining techniques on chromosomes replicated twice in medium containing 5-bromodeoxyuridine (BrdU) and pretreated with Hoechst plus black light. Both silver-nitrate and silver-carbonate staining were affected by chemical extraction and enzyme

Articles

Silver nanomaterials have unique physical, chemical, and optical properties that are currently being leveraged for a wide variety of biological applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service