Skip to Content
MilliporeSigma
All Photos(3)

Documents

851493

Sigma-Aldrich

Maltotriose hydrate

95%

Synonym(s):

O-α-D-Glucopyranosyl-(1→4)-O-α-D-glucopyranosyl-(1→4)-O-α-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O16 · xH2O
CAS Number:
Molecular Weight:
504.44 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

crystals

optical activity

[α]24/D +162°, c = 2 in H2O

mp

132-135 °C (lit.)

SMILES string

[H]O[H].OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)C(O)O[C@@H]3CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H32O16.H2O/c19-1-4-7(22)8(23)12(27)17(31-4)34-15-6(3-21)32-18(13(28)10(15)25)33-14-5(2-20)30-16(29)11(26)9(14)24;/h4-29H,1-3H2;1H2/t4-,5-,6-,7-,8+,9-,10-,11-,12-,13-,14-,15-,16?,17-,18-;/m1./s1

InChI key

HNKASWRMLBJLKJ-HNNWOXMSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Maltotriose is an oligosaccharide with α-1,4 glycoside linkage. It does not possess any ionic groups, hence is suitable for examining the interaction between molecular motion and water molecules.

Application

Maltotriaose hydrate can be used:
  • for sensitive analysis of simple sugars
  • to study heat capacities for sugars and sugar alcohols in dilute aqueous solution by calorimetry
  • as an internal standard in the analysis of sugar levels, during fermentation process in wort samples using HPLC and spectrophotometry

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dielectric relaxation of aqueous solutions of maltotriose
Biorheology, 31, 1620-1620 (2017)
Utilisation of pH stacking in conjunction with a highly absorbing chromophore, 5-aminofluorescein, to improve the sensitivity of capillary electrophoresis for carbohydrate analysis
Journal of Chromatography A, 1200, 8491-8491 (2008)
Monitoring of monosaccharides, oligosaccharides, ethanol and glycerol during wort fermentation by biosensors, HPLC and spectrophotometry
R Monosik
Food Chemistry, 138, 220-226 (2013)
Heat capacities of maltose, maltotriose, maltotetrose and alpha-, beta-, and gamma-cyclodextrin in the solid state and in dilute aqueous solution
LE Briggner
The Journal of Chemical Thermodynamics, 22, 1067-1074 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service