Skip to Content
MilliporeSigma

809012

L-Ornithine-13C5,15N2 hydrochloride

98 atom %, 98% (CP)

Synonym(s):

2,5-Diaminopentanoic-13C5,15N2 acid hydrochloride

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
13C5H1215N2O2 · HCl
CAS Number:
Molecular Weight:
175.57
PubChem Substance ID:
UNSPSC Code:
12352101
Isotopic purity:
98 atom %
Assay:
98% (CP)
Mass shift:
M+7
Form:
powder


isotopic purity

98 atom %

assay

98% (CP)

form

powder

technique(s)

bio NMR: suitable

mass shift

M+7

SMILES string

[15NH2][13CH2][13CH2][13CH2][13C@H]([15NH2])[13C](O)=O.[H]Cl

InChI

1S/C5H12N2O2.ClH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H/t4-;/m0./s1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1;

InChI key

GGTYBZJRPHEQDG-DSNAFDLYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library