Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

807036

Sigma-Aldrich

Silver nanospheres

60 nm avg. part. size, 1 mg/mL (aqueous sodium citrate), citrate functionalized

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141740
NACRES:
NA.23

form

nanospheres

Quality Level

concentration

1 mg/mL (aqueous sodium citrate)

avg. part. size

60 nm

λmax

430 nm

functional group

citrate

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Silver nanomaterials have unique physical, chemical and optical properties that are currently being leveraged for a wide variety of biological applications. Additionally, silver nanoparticles have an optical color that is a function of the silver nanoparticle′s size and shape. The strong coupling of silver nanoparticles to specific wavelengths of incident light can be utilized to develop ultrabright reporter molecules, highly efficient thermal absorbers, and nanoscale “antennas” that can amplify the strength of the local electromagnetic field.

Preparation Note

Shake well before use.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service