Skip to Content
MilliporeSigma
All Photos(1)

Documents

806803

Sigma-Aldrich

Silica

mesoporous SBA-15, <150 μm particle size, pore size 4 nm, Hexagonal pore morphology

Synonym(s):

Porous Silica, SBA-15, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

powder

surface area

750-850 m2/g

particle size

<150 μm

pore size

0.7-0.9 cm3/g pore volume
4 nm pore size

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Mesoporous silicas are inorganic materials having porous structures and find applications in chromatography, catalysis, drug delivery and imaging.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wang Y
Advanced Materials, 273, 576-576 (2015)
Si-Han Wu et al.
Chemical Society reviews, 42(9), 3862-3875 (2013-02-14)
Good control of the morphology, particle size, uniformity and dispersity of mesoporous silica nanoparticles (MSNs) is of increasing importance to their use in catalyst, adsorption, polymer filler, optical devices, bio-imaging, drug delivery, and biomedical applications. This review discusses different synthesis
Isabel Sierra et al.
Journal of chromatography. A, 1363, 27-40 (2014-07-13)
Recent advances in the development of new materials are having a major impact on analytical chemistry. For example, the unique properties of ordered mesoporous silicas (OMSs) have been shown to enhance the analytical performance of many existing techniques or allow

Articles

Mesoporous silica used in wastewater remediation, air cleaning, catalysis, bio-catalysis, and drug delivery.

Mesoporous silica used in wastewater remediation, air cleaning, catalysis, bio-catalysis, and drug delivery.

Mesoporous silica used in wastewater remediation, air cleaning, catalysis, bio-catalysis, and drug delivery.

Mesoporous silica used in wastewater remediation, air cleaning, catalysis, bio-catalysis, and drug delivery.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service