Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

806137

Sigma-Aldrich

bis-MPA-Azide dendrimer

trimethylol propane core, generation 1

Synonym(s):

G1-TMP-Azide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H92N18O18
Molecular Weight:
1317.45
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

liquid

storage temp.

−20°C

SMILES string

O=C(OCC(CC)(COC(C(COC(CCCCCN=[N+]=[N-])=O)(COC(CCCCCN=[N+]=[N-])=O)C)=O)COC(C(COC(CCCCCN=[N+]=[N-])=O)(COC(CCCCCN=[N+]=[N-])=O)C)=O)C(COC(CCCCCN=[N+]=[N-])=O)(COC(CCCCCN=[N+]=[N-])=O)C

InChI

1S/C57H92N18O18/c1-5-57(42-91-51(82)54(2,36-85-45(76)24-12-6-18-30-64-70-58)37-86-46(77)25-13-7-19-31-65-71-59,43-92-52(83)55(3,38-87-47(78)26-14-8-20-32-66-72-60)39-88-48(79)27-15-9-21-33-67-73-61)44-93-53(84)56(4,40-89-49(80)28-16-10-22-34-68-74-62)41-90-50(81)29-17-11-23-35-69-75-63/h5-44H2,1-4H3

InChI key

YTBPIOXKVOSWIN-UHFFFAOYSA-N

General description

  • End Group Functionality: Azide surface group
  • Generation: 1
  • No. Surface Groups: 6
  • Calculated Mol. Wt.: 1317.5 g/mol
  • bis-MPA = 2,2-bis(hydroxymethyl)propionic acid

Application

bis-MPA-Azide dendrimer belongs to the family of nontoxic dendrimers for drug delivery, with functional groups for conjugation or additional modifications.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical Society Reviews, 13, 4593-4617 (2013)
Journal of Polymer Science: Part A, General Papers, 14, 2835-2846 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service