Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

805416

Sigma-Aldrich

5H-Pyrido[3,2-b]indole

Synonym(s):

δ-Carboline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8N2
CAS Number:
Molecular Weight:
168.19
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99% (HPLC)

Quality Level

form

powder

mp

209-214 °C

SMILES string

C1(C=CC=C2)=C2C(N=CC=C3)=C3N1

InChI

1S/C11H8N2/c1-2-5-9-8(4-1)11-10(13-9)6-3-7-12-11/h1-7,13H

InChI key

NSBVOLBUJPCPFH-UHFFFAOYSA-N

Application

Novel aromatic amine compound for organic electroluminescent device, illuminating device and display.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manipulation of electron deficiency of δ-carboline derivatives as bipolar hosts for blue phosphorescent organic light-emitting diodes with high efficiency at 1000 cd m-2.
Zhang Z, et al.
Journal of Material Chemistry C, 4, 4226-4235 (2016)
Spectral and photophysical properties of the δ-carboline anhydrobase (N1-methyl-5H-pyrido[3,2-b]indole).
Balon M, et al.
Chemical Physics, 302(1-3), 13-20 (2004)
Manipulation of electron deficiency of δ-carboline derivatives as bipolar hosts for blue phosphorescent organic light-emitting diodes with high efficiency at 1000 cd m-2.
Zhang Z,et al.
Journal of Material Chemistry C, 4(19), 4226-4235 null
Controlling the exciton lifetime of blue thermally activated delayed fluorescence emitters using a heteroatom-containing pyridoindole donor moiety.
Kim G H, et al.
Materials Horizons, 4, 619-624 (2017)
Thermally activated delayed fluorescence of co-deposited copper(i) complexes: cost-effective emitters for highly efficient organic light-emitting diodes.
Wang Z, et al.
Journal of Material Chemistry C, 5, 6982-6988 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service