Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

805033

Sigma-Aldrich

Carbon nanotube, single-walled

carbon ≥80 %, diam. 1.2-2.0 nm, avg. no. of layers, 1

Synonym(s):

CNT, SWCNT, SWNT, Single wall carbon nanotubes, Tuball

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

powder

Quality Level

feature

avg. no. of layers 1

composition

carbon, ≥80%

L

>5 μm , AFM

diam.

1.2-2.0 nm

surface area

520 m2/g

average diameter

1.8 nm

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Application

Used as additives.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Single-Walled Carbon Nanotubes synthesized by the Super-Growth Method & their properties & applications, including dispersing SGCNTs, SGCNT-polymer composites & SGCNT-metal composites are discussed.

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries.

Boron nitride nanotubes (BNNT) are close structural analogs of carbon nanotubes (CNT), which are high aspect ratio nanotubular material, where carbon atoms are alternately substituted by nitrogen and boron atoms.

Boron nitride nanotubes (BNNT) are close structural analogs of carbon nanotubes (CNT), which are high aspect ratio nanotubular material, where carbon atoms are alternately substituted by nitrogen and boron atoms.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service