Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

799238

Sigma-Aldrich

H-Lys(Boc)-OH-13C6,15N2

99 atom % 13C, 98 atom % 15N, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C5H2215N2O4
CAS Number:
Molecular Weight:
254.25
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % 15N

Assay

95% (CP)

form

powder

technique(s)

bio NMR: suitable

application(s)

peptide synthesis

functional group

Boc

mass shift

M+8

storage temp.

2-8°C

SMILES string

O[13C]([13C@]([H])([15NH2])[13CH2][13CH2][13CH2][13CH2][15NH]C(OC(C)(C)C)=O)=O

InChI

1S/C11H22N2O4/c1-11(2,3)17-10(16)13-7-5-4-6-8(12)9(14)15/h8H,4-7,12H2,1-3H3,(H,13,16)(H,14,15)/t8-/m1/s1/i4+1,5+1,6+1,7+1,8+1,9+1,12+1,13+1

InChI key

VVQIIIAZJXTLRE-JHLHIYTKSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service