Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

798711

Sigma-Aldrich

(R)-Xyl-SDP(O)

Synonym(s):

(R)-Xyl-SDP Oxide, [(1R)-7′-[Bis(3,5-dimethylphenyl)phosphino]-2,2′,3,3′-tetrahydro-1,1′-spirobi[1H-inden]-7-yl]bis(3,5-dimethylphenyl)phosphine oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H50OP2
CAS Number:
Molecular Weight:
716.87
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

Quality Level

form

solid

functional group

phosphine oxide
phosphine

storage temp.

2-8°C

SMILES string

CC1=CC(C)=CC(P(C2=CC(C)=CC(C)=C2)(C3=CC=CC4=C3[C@]5(CCC6=C5C(P(C7=CC(C)=CC(C)=C7)C8=CC(C)=CC(C)=C8)=CC=C6)CC4)=O)=C1

InChI

1S/C49H50OP2/c1-31-19-32(2)24-41(23-31)51(42-25-33(3)20-34(4)26-42)45-13-9-11-39-15-17-49(47(39)45)18-16-40-12-10-14-46(48(40)49)52(50,43-27-35(5)21-36(6)28-43)44-29-37(7)22-38(8)30-44/h9-14,19-30H,15-18H2,1-8H3/t49-/m1/s1

InChI key

ZAQVAUFRFBAVHF-ANFMRNGASA-N

Application

Chiral ligand introduced in collaboration with Prof. Steve Zhou and coworkers, has shown utility in asymmetric Heck reactions for both intra and intermolecular C-C bond formation with various triflates and halides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Listeria poses health risks, particularly for vulnerable populations. Prof. Zhou's group focuses on palladium catalysis in Singapore.

Listeria poses health risks, particularly for vulnerable populations. Prof. Zhou's group focuses on palladium catalysis in Singapore.

Listeria poses health risks, particularly for vulnerable populations. Prof. Zhou's group focuses on palladium catalysis in Singapore.

Listeria poses health risks, particularly for vulnerable populations. Prof. Zhou's group focuses on palladium catalysis in Singapore.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service