Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

796875

Sigma-Aldrich

Fluoroacetic acid

95%

Synonym(s):

α-Fluoroacetic acid, Cymonic acid, Fluoroethanoic acid, Gifblaar poison, Monofluoroacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H3FO2
CAS Number:
Molecular Weight:
78.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

functional group

carboxylic acid
fluoro

SMILES string

O=C(O)CF

InChI

1S/C2H3FO2/c3-1-2(4)5/h1H2,(H,4,5)

InChI key

QEWYKACRFQMRMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhi-Qiang Liu et al.
Pharmacology, biochemistry, and behavior, 143, 44-56 (2016-02-11)
D-serine, the endogenous co-agonist of N-methyl-D-aspartate receptors (NMDARs), is considered to be essential for learning and memory. The aim of the current investigation was to systematically evaluate the role of D-serine on addiction behaviors considered to be mediated by the
E V Sidorin et al.
Biochemistry. Biokhimiia, 82(11), 1304-1313 (2017-12-11)
Recombinant porin OmpF (an integral protein of bacterial outer membrane) from Yersinia pseudotuberculosis was synthesized in Escherichia coli cells as inclusion bodies. By combining the methods of anion-exchange and gel filtration chromatographies, recombinant OmpF (rOmpF) was isolated as an individual
Sebastian Schneider et al.
Plant, cell & environment, 42(4), 1180-1189 (2018-11-18)
Legume-rhizobia symbioses play a major role in food production for an ever growing human population. In this symbiosis, dinitrogen is reduced ("fixed") to ammonia by the rhizobial nitrogenase enzyme complex and is secreted to the plant host cells, whereas dicarboxylic
Miao Dong et al.
Fish & shellfish immunology, 70, 260-269 (2017-09-09)
Growing evidence suggests that the immune system of teleost is vulnerable to xenoestrogens, which are ubiquitous in the marine environment. This study detected and identified the major circulatory immune proteins deregulated by 17α-ethinylestradiol (EE2), which may be linked to fish
Juan Zhang et al.
Soft matter, 13(3), 670-676 (2016-12-24)
From catanionic fluoro-/hydro-carbon mixtures of fluoroacetic acid (CF

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service