Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

796549

Sigma-Aldrich

Stahl Aerobic Oxidation TEMPO solution

0.2 M in acetonitrile, Solution for Oxidation of Primary Alcohols

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

Quality Level

form

liquid

reaction suitability

reagent type: oxidant

concentration

0.2 M in acetonitrile

storage temp.

2-8°C

SMILES string

CN1C=CN=C1.CC2(C)CCCC(C)(C)N2[O].C3(C4=NC=CC=C4)=NC=CC=C3

InChI

1S/C10H8N2.C9H18NO.C4H6N2/c1-3-7-11-9(5-1)10-6-2-4-8-12-10;1-8(2)6-5-7-9(3,4)10(8)11;1-6-3-2-5-4-6/h1-8H;5-7H2,1-4H3;2-4H,1H3

InChI key

BQFURWVGIDXRNB-UHFFFAOYSA-N

General description

Stahl Aerobic Oxidation TEMPO solution contains 1-methylimidazole (NMI), 2,2′-bipyridyl (bpy), and TEMPO. It is widely employed for the oxidation of alcohols.

Application

This convenience solution can be used in tandem with tetrakisacetonitrile copper(I) triflate (685038) for the oxidation of primary alcohols through aerobic conditions developed by the Stahl Research Group.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1C

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F

Flash Point(C)

2.0 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Continuous-Flow Aerobic Oxidation of Primary Alcohols with a Copper(I)/TEMPO Catalyst
Greene, JF, et al.
Organic Process Research & Development, 17 (10), 1247?1251-1247?1251 (2013)
Practical Aerobic Alcohol Oxidation with Cu/Nitroxyl and Nitroxyl/NOx Catalyst Systems.
Miles KC and Stahl SS.
Aldrichimica Acta, 48(1) (2015)
Jessica M Hoover et al.
Journal of the American Chemical Society, 133(42), 16901-16910 (2011-08-25)
Aerobic oxidation reactions have been the focus of considerable attention, but their use in mainstream organic chemistry has been constrained by limitations in their synthetic scope and by practical factors, such as the use of pure O(2) as the oxidant
Jessica M Hoover et al.
Nature protocols, 7(6), 1161-1166 (2012-05-29)
This protocol describes a practical laboratory-scale method for aerobic oxidation of primary alcohols to aldehydes, using a chemoselective Cu(I)/TEMPO (TEMPO = 2,2,6,6-tetramethyl-1-piperidinyloxyl) catalyst system. The catalyst is prepared in situ from commercially available reagents, and the reactions are performed in
Janelle E Steves et al.
The Journal of organic chemistry, 80(21), 11184-11188 (2015-10-13)
Two solutions, one consisting of bpy/TEMPO/NMI and the other bpy/ABNO/NMI (bpy =2,2'-bipyridyl; TEMPO = 2,2,6,6-tetramethylpiperidine N-oxyl, ABNO = 9-azabicyclo[3.3.1]nonane N-oxyl; NMI = N-methylimidazole), in acetonitrile are shown to have good long-term stability (≥1 year) under air at 5 °C. The

Articles

Alcohol oxidation yields aldehydes and ketones, crucial intermediates in organic synthesis.

Alcohol oxidation yields aldehydes and ketones, crucial intermediates in organic synthesis.

Alcohol oxidation yields aldehydes and ketones, crucial intermediates in organic synthesis.

Alcohol oxidation yields aldehydes and ketones, crucial intermediates in organic synthesis.

Related Content

he Stahl Lab focuses on the development of catalysts and catalytic reactions for selective oxidation of organic molecules, with particular emphasis on aerobic oxidation reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service