Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

79412

Sigma-Aldrich

Phosphazene base P1-t-Oct

≥97.0%

Synonym(s):

tert-Octylimino-tris(dimethylamino)phosphorane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H35N4P
CAS Number:
Molecular Weight:
290.43
Beilstein:
7129981
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

form

liquid

refractive index

n20/D 1.474

bp

68 °C/0.05 mmHg (lit.)

functional group

amine

SMILES string

CN(C)P(=NC(C)(C)CC(C)(C)C)(N(C)C)N(C)C

InChI

1S/C14H35N4P/c1-13(2,3)12-14(4,5)15-19(16(6)7,17(8)9)18(10)11/h12H2,1-11H3

InChI key

DSCJCKAURXOQPX-UHFFFAOYSA-N

Other Notes

Sterically hindered, neutral nitrogen base with similar basicity as BEMP (Cat. No. 20025)

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Schwesinger et al
Angewandte Chemie (International Edition in English), 105, 1420-1420 (1993)

Articles

Phosphazene base reagents are available as monomeric (P1 and BEMP), dimeric (P2), and tetrameric (P4) bases with different side chains to control their sterical hindrance.

Phosphazene base reagents are available as monomeric (P1 and BEMP), dimeric (P2), and tetrameric (P4) bases with different side chains to control their sterical hindrance.

Phosphazene base reagents are available as monomeric (P1 and BEMP), dimeric (P2), and tetrameric (P4) bases with different side chains to control their sterical hindrance.

Phosphazene base reagents are available as monomeric (P1 and BEMP), dimeric (P2), and tetrameric (P4) bases with different side chains to control their sterical hindrance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service