Skip to Content
MilliporeSigma
All Photos(1)

Documents

793485

Sigma-Aldrich

Melengestrol acetate-d3

≥98 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25D3H29O4
Molecular Weight:
399.54
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

≥98 atom % D

Assay

≥98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

storage temp.

2-8°C

SMILES string

C=C1C[C@@]2([H])[C@]3([H])C=C(C)C4=CC(CC[C@]4(C)[C@@]3([H])CC[C@]2(C)[C@]1(C(C)=O)OC(C([2H])([2H])[2H])=O)=O

InChI

1S/C25H32O4/c1-14-11-19-20(23(5)9-7-18(28)13-21(14)23)8-10-24(6)22(19)12-15(2)25(24,16(3)26)29-17(4)27/h11,13,19-20,22H,2,7-10,12H2,1,3-6H3/t19-,20+,22+,23-,24+,25+/m1/s1/i4D3

InChI key

UDKABVSQKJNZBH-TWOXOXTASA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brett R Blackwell et al.
Environmental science & technology, 49(14), 8796-8803 (2015-06-23)
Studies of steroid growth promoters from beef cattle feedyards have previously focused on effluent or surface runoff as the primary route of transport from animal feeding operations. There is potential for steroid transport via fugitive airborne particulate matter (PM) from
Shuang-Shuang Liu et al.
Analytical and bioanalytical chemistry, 406(28), 7299-7311 (2014-09-28)
In this study, a highly sensitive and robust method using an ultra-high-performance liquid chromatography-tandem mass spectrometry combined with solid-phase extraction and ultrasonic extraction for pretreatment and silica gel purification steps has been developed for determination of 21 natural and synthetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service