Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

792446

Sigma-Aldrich

Sodium 1,1-difluoro-4-(2-methyl-1,3-dioxolan-2-yl)butane-1-sulfinate

Synonym(s):

Shabat Sulfinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13F2NaO4S
CAS Number:
Molecular Weight:
266.24
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: C-C Bond Formation
reagent type: catalyst
reaction type: C-H Activation

reagent type: linker

SMILES string

[O-]S(C(F)(F)CCCC1(C)OCCO1)=O.[Na+]

InChI

1S/C8H14F2O4S.Na/c1-7(13-5-6-14-7)3-2-4-8(9,10)15(11)12;/h2-6H2,1H3,(H,11,12);/q;+1/p-1

InChI key

UDXMRVSKBPZBMK-UHFFFAOYSA-M

Application

Shabat sulfinate is a difluoroalkyl ketal sulfinate sodium salt that has been used as a reagent for the introduction of the ketone functional groups on heterocyclic bioactive molecules via derivatization of C-H bonds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samer Gnaim et al.
Bioconjugate chemistry, 27(9), 1965-1971 (2016-08-06)
We have developed a new difluoroalkyl ketal sulfinate salt reagent suitable for direct derivatization of heteroarene C-H bonds. The reagent is capable of introducing a ketone functional group on heteroarene bioactive compounds via a one-pot reaction. Remarkably, in three examples

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service