Skip to Content
MilliporeSigma
All Photos(1)

Documents

791881

Sigma-Aldrich

Conductive silver printing ink, resistivity 9 - 10 μΩ-cm

Synonym(s):

Greatcell Solar®, Dyesol® DYAG100 conductive silver ink, Screen printable silver paste

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

description

volume resistivity 9 - 10 μΩ-cm

Quality Level

Assay

75-85% (Solids content)

form

paste (white)

viscosity

9000-12000 mPa.s (at shear rate of 10 sec-1 at 25°C)

General description

Sheet Resistivity is 3.6 - 4 mΩ / square / mil. (at a 180°C cure temperature)

Application

Use this Conductive Silver Ink to achieve outstanding conductivity at low cure temperatures when printing on a variety of substrates, including glass and polyesters (PET/PEN) and ITO/FTO coated substrates.

This Conductive Silver Printing Ink provides outstanding conductivity at low cure temperatures. It provides extremely low resistance of printed conducting tracks, essential for the photovoltaic applications, and is a key enabler in plastic electronics such as DSSC; OPV; CdTe solar cells etc.,by providing lowest track resistances of printed current collectors.

The Conductive Silver Printing Ink is comprised of a mixture of pure silver particles and an organometallic silver compound in an organic medium. When printed on polyester films, and then heated to 160 - 180°C, the particles consolidate to form a chemically-welded, continuous metal track.

Legal Information

Product of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar Materials Pty Ltd.
Dyesol is a registered trademark of Greatcell Solar
Greatcell Solar is a registered trademark of Greatcell Solar

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F

Flash Point(C)

79 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The ability to pattern conductive electrodes is technologically relevant for several applications, including photovolatics, displays, sensors, and biomedical devices.

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service