Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

790575

Sigma-Aldrich

(2-Boc-amino)ethyl methacrylate

99% (GC)

Synonym(s):

2-(N-tert-Butoxycarbonylamino)ethyl methacrylate, 2-(tert-Butoxycarbonylamino)ethyl methacrylate, 2-Methyl-2-propenoic acid 2-[[(1,1-dimethylethoxy)carbonyl]amino]ethyl ester, BOC protected aminoethyl methacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO4
CAS Number:
Molecular Weight:
229.27
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99% (GC)

form

solid

mp

78-84 °C

SMILES string

C=C(C)C(OCCNC(OC(C)(C)C)=O)=O

InChI

1S/C11H19NO4/c1-8(2)9(13)15-7-6-12-10(14)16-11(3,4)5/h1,6-7H2,2-5H3,(H,12,14)

InChI key

HZWCSJMONGVKJV-UHFFFAOYSA-N

Application

The methacrylate can be polymerized to generate a polymer with pendant amine functionality. The protecting group, BOC, is usually removed with acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service