Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

779660

Sigma-Aldrich

γ-Cyclodextrin

Wacker Chemie AG, 98.3-102.0% cyclodextrin basis

Synonym(s):

Cavamax® W8, Cyclomaltooctaose, Cyclooctaamylose, Schardinger γ-Dextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H80O40
CAS Number:
Molecular Weight:
1297.12
Beilstein:
78740
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98.3-102.0% cyclodextrin basis

form

crystals

optical activity

[α]/D 174.0 to 180.0°

manufacturer/tradename

Wacker Chemie AG

impurities

≤0.20% reducing substances
≤0.5% related substances (sum)
≤0.50% impurities A, B and C (related substances) (single)
≤5 ppm heavy metals
≤8 ppm volatile organics

ign. residue

≤0.10%

loss

≤11.0% loss on drying

λ

10 % in H2O

UV absorption

λ: 420 nm Amax: ≤0.20

functional group

ether
hydroxyl

SMILES string

[H][C@]1(O[C@H](COC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@H]1OC(C)=O)O[C@@H]2[C@@H](COC(C)=O)O[C@H](OC(C)=O)[C@H](OC(C)=O)[C@H]2OC(C)=O

InChI

1S/C48H80O40/c49-1-9-33-17(57)25(65)41(73-9)82-34-10(2-50)75-43(27(67)19(34)59)84-36-12(4-52)77-45(29(69)21(36)61)86-38-14(6-54)79-47(31(71)23(38)63)88-40-16(8-56)80-48(32(72)24(40)64)87-39-15(7-55)78-46(30(70)22(39)62)85-37-13(5-53)76-44(28(68)20(37)60)83-35-11(3-51)74-42(81-33)26(66)18(35)58/h9-72H,1-8H2/t9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48-/m1/s1

InChI key

GDSRMADSINPKSL-HSEONFRVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

γ-Cyclodextrin (γ-CD), a water-soluble cyclic oligosaccharide consisting of eight α-(1,4)-linked glucopyranose subunits, can be used in a wide range of applications including:
  • Synthesis of renewable and biocompatible metal-organic frameworks.
  • Formation of inclusion complexes with a variety of guest molecules to increase their solubility in water and other polar solvents.
  • Synthesis of γ-CD based nanogels and dendrimers as carriers and stabilizers for drug delivery applications.

Legal Information

Cavamax is a registered trademark of Wacker Chemie AG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclodextrin-mediated enhancement of riboflavin solubility and corneal permeability.
Morrison, Peter WJ et al.
Molecular Pharmaceutics, 10(2), 756-762 (2013)
Cross-linked hydroxypropyl-β-cyclodextrin and γ-cyclodextrin nanogels for drug delivery: physicochemical and loading/release properties.
Moya-Ortega, Maria D et al.
Carbohydrate Polymers, 87(3), 2344-2351 (2012)
Metal-organic frameworks from edible natural products.
Smaldone, Ronald A et al.
Angewandte Chemie (International Edition in English), 49(46), 8630-8634 (2010)
γ-Cyclodextrin: a review on enzymatic production and applications.
Li, Zhaofeng et al.
Applied Microbiology and Biotechnology, 77(2), 245-245 (2007)
Cyclodextrin functionalized graphene nanosheets with high supramolecular recognition capability: synthesis and host- guest inclusion for enhanced electrochemical performance.
Guo, Yujing et al.
ACS Nano, 4(7), 4001-4010 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service