Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

778907

Sigma-Aldrich

(2-Hydroxypropyl)-β-cyclodextrin

Produced by Wacker Chemie AG, Burghausen, Germany, Life Science

Synonym(s):

Cavasol® W7 HP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352201
NACRES:
NA.22

grade

Produced by Wacker Chemie AG, Burghausen, Germany, Life Science

Quality Level

form

powder

extent of labeling

0.59-0.73 molar substitution

impurities

≤0.1% related substances, any (Ph Eur)
≤0.5% propylene glycol (Ph Eur)
≤1.0% β-dex (Ph Eur)
≤1.0% related substances, total other
≤1.0 ppm propylene oxide

loss

≤10.0% loss on drying

conductivity

≤200 μS/cm (Ph Eur)

suitability

clear, colorless for appearance of solution
positive for identity

SMILES string

CC(O)COCC1OC2OC3C(COCC(C)O)OC(OC4C(COCC(C)O)OC(OC5C(COCC(C)O)OC(OC6C(COCC(C)O)OC(OC7C(COCC(C)O)OC(OC8C(COCC(C)O)OC(OC1C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C8OCC(C)O)C(OCC(C)O)C7OCC(C)O)C(OCC(C)O)C6OCC(C)O)C(OCC(C)O)C5OCC(C)O)C(OCC(C)O)C4OCC(C)O)C(OCC(C)O)C3OCC(C)O

InChI

1S/C63H112O42/c1-22(64)8-85-15-29-50-36(71)43(78)57(92-29)100-51-30(16-86-9-23(2)65)94-59(45(80)38(51)73)102-53-32(18-88-11-25(4)67)96-61(47(82)40(53)75)104-55-34(20-90-13-27(6)69)98-63(49(84)42(55)77)105-56-35(21-91-14-28(7)70)97-62(48(83)41(56)76)103-54-33(19-89-12-26(5)68)95-60(46(81)39(54)74)101-52-31(17-87-10-24(3)66)93-58(99-50)44(79)37(52)72/h22-84H,8-21H2,1-7H3/t22?,23?,24?,25?,26?,27?,28?,29-,30-,31?,32?,33?,34?,35?,36?,37-,38?,39-,40+,41+,42+,43?,44+,45?,46+,47+,48+,49+,50+,51+,52-,53-,54-,55-,56-,57+,58-,59+,60-,61-,62-,63-/m1/s1

InChI key

ODLHGICHYURWBS-RYJYQAAZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(2-Hydroxypropyl)-β-cyclodextrin can be used:
  • In the preparation of imidazole nucleosides via enzymatic transglycosylation to improve the solubility and/or stability of 4-aryl and 4-heteroaryl-1H-imidazole reactants.
  • Cyanazine-hydroxypropyl-β-cyclodextrin inclusion complex with enhanced water solubility when compared to cyanazine.

The solubility of lipophilic drugs increases linearly with the concentration of hydroxypropyl-β-cyclodextrin (HBC) in aqueous solution because of the complex between HBC and the drug. This guest-host type complex is formed between the drug and the non-polar cavity in the HBC that results in enhanced solubility. Solutions may be lyophilized to produce freely soluble powders. Non-toxic in rabbits and mice.

Legal Information

Cavasol is a registered trademark of Wacker Chemie AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expedient and generic synthesis of imidazole nucleosides by enzymatic transglycosylation
Vichier-Guerre S, et al.
Organic & Biomolecular Chemistry, 14(14), 3638-3653 (2016)
Preparation and characterization of cyanazine-hydroxypropyl-β-cyclodextrin inclusion complex
Gao S, et al.
Royal Society of Chemistry Advances, 9(45), 26109-26115 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service