Skip to Content
MilliporeSigma
All Photos(1)

Documents

778508

Sigma-Aldrich

4-Hexylphenol

≥98.0% (HPLC)

Synonym(s):

1-(4-Hydroxyphenyl)hexane, p-n-Hexylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18O
CAS Number:
Molecular Weight:
178.27
Beilstein:
387122
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

crystals

mp

23-32 °C (lit.)

SMILES string

CCCCCCc1ccc(O)cc1

InChI

1S/C12H18O/c1-2-3-4-5-6-11-7-9-12(13)10-8-11/h7-10,13H,2-6H2,1H3

InChI key

SZWBRVPZWJYIHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Pastor-Belda et al.
Food chemistry, 221, 76-81 (2016-12-17)
Magnetic solid phase extraction (MSPE) with cobalt ferrite nanoparticles coated with oleic acid is described for the determination of alkylphenols (APs), 4-tert-butylphenol (TBP), 4-pentylphenol (PP), 4-hexylphenol (HP), 4-tert-octylphenol (TOP), 4-n-octylphenol (OP) and 4-nonylphenol (NP) in baby foods using gas chromatography

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service