Skip to Content
MilliporeSigma
All Photos(3)

Documents

778222

Sigma-Aldrich

Boc-D-Lys-OH

98%

Synonym(s):

Nα-(tert-Butoxycarbonyl)-D-lysine, Nα-Boc-D-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22N2O4
CAS Number:
Molecular Weight:
246.30
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder or crystals

optical activity

[α]22/D -24°, c = 0.5% in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

205-210 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

OC([C@H](NC(OC(C)(C)C)=O)CCCCN)=O

InChI

1S/C11H22N2O4/c1-11(2,3)17-10(16)13-8(9(14)15)6-4-5-7-12/h8H,4-7,12H2,1-3H3,(H,13,16)(H,14,15)/t8-/m1/s1

InChI key

DQUHYEDEGRNAFO-MRVPVSSYSA-N

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service