Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

772836

Sigma-Aldrich

Isopropenylmagnesium bromide solution

greener alternative

1.0 M in 2-methyltetrahydrofuran

Synonym(s):

1-Methylvinylmagnesium bromide, 1-Propen-2-ylmagnesium bromide, 2-Propen-2-ylmagnesium bromide, Bromo prop-1-en-2-yl magnesium, Bromo(isopropenylmagnesium), Isopropenylmagnesium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)MgBr
CAS Number:
Molecular Weight:
145.28
Beilstein:
3587297
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
EC Index Number:
603-688-6
NACRES:
NA.22

form

solution

Quality Level

reaction suitability

reaction type: Grignard Reaction

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

1.0 M in 2-methyltetrahydrofuran

density

0.970 g/mL

greener alternative category

storage temp.

2-8°C

SMILES string

CC(=C)[Mg]Br

InChI

1S/C3H5.BrH.Mg/c1-3-2;;/h1H2,2H3;1H;/q;;+1/p-1

InChI key

LFTYALBVGVNGLI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product, 1.0 M in 2-methyltetrahydrofuran aligns with Safer Solvents and Auxiliaries, Use of Renewable Feedstocks and Inherently Safer Chemistry for Accident Prevention. Click here for more information.

Application

Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF)

2-Methyltetrahydrofuran (2-MeTHF): A Biomass-Derived Solvent with Broad Application in Organic Chemistry

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service