Skip to Content

772011

Fmoc-D-Asp(OtBu)-OH

98%

Synonym(s):

Fmoc-D-aspartic acid 4-tert-butyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H25NO6
CAS Number:
Molecular Weight:
411.45
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

98%

form

powder or crystals

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

color

white to beige

mp

146-151 °C

application(s)

peptide synthesis

functional group

Fmoc

SMILES string

N([C@H](CC(=O)OC(C)(C)C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C23H25NO6/c1-23(2,3)30-20(25)12-19(21(26)27)24-22(28)29-13-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,18-19H,12-13H2,1-3H3,(H,24,28)(H,26,27)/t19-/m1/s1

InChI key

FODJWPHPWBKDON-LJQANCHMSA-N



Still not finding the right product?

Explore all of our products under Fmoc-D-Asp(OtBu)-OH


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library