Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

769932

Sigma-Aldrich

Resomer® L 209 S, Poly(L-lactide)

ester terminated

Synonym(s):

Resomer® L 209 S, PLLA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

form

granular

viscosity

2.6-3.2 dL/g

transition temp

Tg 60-65 °C

storage temp.

2-8°C

SMILES string

O1[C@H](C(=O)O[C@H](C1=O)C)C

InChI

1S/C6H8O4/c1-3-5(7)10-4(2)6(8)9-3/h3-4H,1-2H3/t3-,4-/m0/s1

InChI key

JJTUDXZGHPGLLC-IMJSIDKUSA-N

Application

Biodegradable, biocompatible, and bioresorbable polymer composed of L-Lactide. This semi-crystalline material has been used in the fabrication of research medical devices and research tissue engineering solutions, such as orthopedic or soft tissue fixation devices. Degradation of this materials has been thoroughly studied and has been shown to be safely resorbed by the body after implantation. Modification of molecular weight and polymer composition allows for control of the degradation rate and mechanical stability of the polymer.

Legal Information

RESOMER is a registered trademark of Evonik Rohm GmbH

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service