Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

769371

Sigma-Aldrich

Titanium(IV) oxide

pellets, diam. × thickness 12.5 mm × 5.5 mm

Synonym(s):

Titania, Titanium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
EC Number:
MDL number:
UNSPSC Code:
11181502
NACRES:
NA.23

form

pellets

Quality Level

diam. × thickness

12.5 mm × 5.5 mm

impurities

≤0.1% PBB (polybrominated biphenyls)
≤0.1% PBDE (polybrominated diphenyl ethers)

color

black

mp

>350 °C (lit.)

density

4.26 g/mL at 25 °C (lit.)

cation traces

Cd: ≤0.01%
Co: ≤0.001%
Cr: ≤0.002%
Cu: ≤0.001%
Fe: ≤0.01%
Hg: ≤0.1%
Pb: ≤0.1%
V: ≤0.005%

SMILES string

O=[Ti]=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Black titanium dioxide (TiO2) contains surface disorders in the nanophase TiO2 through hydrogenation, resulting in a lattice disorder on the surface. This material is a widely studied photocatalyst due to its unique properties, such as a low band gap and absorbance of visible light.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

M Barua et al.
Poultry science, 99(12), 6925-6934 (2020-11-30)
The aim of present study was to determine the influence of the feed form (FF) on standardized ileal digestibility (SID) of nitrogen (N) and amino acids (AA) in 3 protein sources (PS) for broiler chickens. Six diets were tested in
Titanium dioxide thin films: preparation and optical properties
Bedikyan , L., et al.
Journal of Chemical Technology and Metallurgy, 48(6), 555-558 (2013)
Titanium dioxide photocatalysis
Fujishima, A., et al.
Journal of Photochemistry and Photobiology, C: Photochemistry Reviews, 1(1), 1-21 (2000)
Fabrication of hydrogen sensors with transparent titanium oxide nanotube-array thin films as sensing elements
Mor, GK., et al.
Thin Solid Films, 496(1), 42-48 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service