Skip to Content
MilliporeSigma
All Photos(3)

Documents

768375

Sigma-Aldrich

Azobisisobutyronitrile

12 wt. % in acetone

Synonym(s):

2,2′-Azobis(2-methylpropionitrile) solution, radical initiator, α,α,′-Azoisobutyronitrile solution, AIBN solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N4
CAS Number:
Molecular Weight:
164.21
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

liquid

concentration

12 wt. % in acetone

refractive index

n20/D 1.368

density

0.808 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)(\N=N\C(C)(C)C#N)C#N

InChI

1S/C8H12N4/c1-7(2,5-9)11-12-8(3,4)6-10/h1-4H3/b12-11+

InChI key

OZAIFHULBGXAKX-VAWYXSNFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Azobisisobutyronitrile (AIBN) is an azo-compound and is widely used as a free radical initiator. This compound has labile carbon-nitrogen covalent bond which undergoes homolytic scission under thermal, chemical or photochemical conditions producing free radicals. They are useful in many reactions like halogenation, polymerisation of vinyl monomers, grafting reactions, curing of rubbers and unsaturated polymers and cross-linking of polyolefins.

Application

  • Used as an initiator in the synthesis of highly cross-linked Poly(divinylbenzene) (PDVB) polymers.
  • Used as an initiator in the polymerization process of 2-hydroxyethyl methacrylate (HEMA).

Features and Benefits

Decomposes unimolecularly at good rates without much variation from one solvent to another.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Initiator efficiency in radical polymerization
Walling, C.
Journal of Polymer Science, 14(74), 214-217 (1954)
Synthesis of highly cross-linked polymers in supercritical carbon dioxide by heterogeneous polymerization.
Cooper, A. I., Hems, W. P., & Holmes, A. B.
Macromolecules, 32(7), 2156-2166 (1999)
Biomaterials based on 2-hydroxyethyl methacrylate: the influence of the initiator type
Nita, L. E., Chiriac, A. P., Nistor, M. T., & Stoica, I.
Rev. Roum. Chim., 56(5), 537-543 (2011)
Selected radical azoinitiators in the synthesis of solvent-borne acrylic pressure-sensitive adhesives
Pabin-Szafko, B., Wisniewska, E., & Czech, Z.
Chemistry and Chemical Technology, 3(2), 101-106 (2009)
Idil Ipek Yilmaz et al.
Macromolecular rapid communications, 33(9), 856-862 (2012-04-21)
Polymers containing maleimide groups on their side chains have been synthesized by utilization of a novel styrenic monomer containing a masked-maleimide unit. AIBN initiated free radical polymerization and reverse addition-fragmentation chain transfer (RAFT) polymerization was utilized for synthesis of copolymers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service