Skip to Content
MilliporeSigma

767751

2-{2-[2-(2-Mercaptoethoxy)ethoxy]ethoxy}ethanol

97%

Synonym(s):

MEEE, 1-Mercapto-11-hydroxy-3,6,9-trioxaundecane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H18O4S
Molecular Weight:
210.29
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
97%
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

97%

form

liquid

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.481

density

1.098 g/mL at 25 °C

SMILES string

OCCOCCOCCOCCS

InChI

1S/C8H18O4S/c9-1-2-10-3-4-11-5-6-12-7-8-13/h9,13H,1-8H2

InChI key

GKKYNULPAQDGHI-UHFFFAOYSA-N

Application

2-{2-[2-(2-Mercaptoethoxy)ethoxy]ethoxy}ethanol may be used in the synthesis of 2-(2-(2-((7-nitrobenzo[c][1,2,5]oxadiazol-4-yl)thio)ethoxy)ethoxy)ethanol, an nitrobenzoxadiazole (NBD) analog of the anticancer agent 6-((7-nitrobenzo[c][1,2,5]oxadiazol-4-yl)thio)hexan-1-ol (NBDHEX). This analog is a potent glutathione S-transferase (GST) and a potential therapeutic antimelanoma agent with enhanced water solubility and antitumor efficacy when compared to NBDHEX in a study.
  • Oligo-ehtylene glycol of peptides and small molecules (PEGylation)
  • PEG-disulfide formation
  • ultra-thin protein-resistant monolayers
  • Densely packed monolayer and a flexible-hydrophilic oligo ethylene glycol arm for avoiding non-specific adsorption


Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library