Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

766682

Sigma-Aldrich

2,2,6,6-Tetramethyl-4-[1-oxo-6-(triethylammonio)hexylamino]-1-piperidinyloxy bromide

95%

Synonym(s):

WS-TEMPO, water soluble N-oxyl compound, water soluble-TEMPO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H43BrN3O2
CAS Number:
Molecular Weight:
449.49
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

reagent type: oxidant

mp

216-221 °C (decomposition)

functional group

amide
amine

storage temp.

2-8°C

SMILES string

[Br-].CC[N+](CC)(CC)CCCCCC(=O)NC1CC(C)(C)N([O])C(C)(C)C1

InChI

1S/C21H42N3O2.BrH/c1-8-24(9-2,10-3)15-13-11-12-14-19(25)22-18-16-20(4,5)23(26)21(6,7)17-18;/h18H,8-17H2,1-7H3;1H

InChI key

NEQWUIVYFXHUMV-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service