Skip to Content
MilliporeSigma
All Photos(2)

Documents

764094

Sigma-Aldrich

1,8-Dibromonaphthalene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6Br2
CAS Number:
Molecular Weight:
285.96
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder or crystals

mp

105-110 °C

storage temp.

2-8°C

SMILES string

Brc1cccc2cccc(Br)c12

InChI

1S/C10H6Br2/c11-8-5-1-3-7-4-2-6-9(12)10(7)8/h1-6H

InChI key

DLXBGTIGAIESIG-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service