Skip to Content
MilliporeSigma
All Photos(3)

Documents

763489

Sigma-Aldrich

5-Fluoro-2-iodoaniline

97%

Synonym(s):

5-Fluoro-2-iodobenzenamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5FIN
CAS Number:
Molecular Weight:
237.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

41-45 °C

storage temp.

2-8°C

SMILES string

Nc1cc(F)ccc1I

InChI

1S/C6H5FIN/c7-4-1-2-5(8)6(9)3-4/h1-3H,9H2

InChI key

UCPDOOZBROQHME-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service