Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

761907

Sigma-Aldrich

4-Bromo-6-iodoquinoline

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5BrIN
Molecular Weight:
333.95
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

powder

mp

173-178 °C

functional group

bromo
iodo

storage temp.

2-8°C

SMILES string

Brc1ccnc2ccc(I)cc12

InChI

1S/C9H5BrIN/c10-8-3-4-12-9-2-1-6(11)5-7(8)9/h1-5H

InChI key

RIMFFKOKLMUJLR-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service