Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

760080

Sigma-Aldrich

4′-Mercaptobiphenylcarbonitrile

97% (GC)

Synonym(s):

4′-Mercapto-[1,1′-biphenyl]-4-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9NS
CAS Number:
Molecular Weight:
211.28
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97% (GC)

form

powder

mp

127-132 °C

SMILES string

Sc1ccc(cc1)-c2ccc(cc2)C#N

InChI

1S/C13H9NS/c14-9-10-1-3-11(4-2-10)12-5-7-13(15)8-6-12/h1-8,15H

InChI key

ZZSHJNKMDIZSFY-UHFFFAOYSA-N

Application

This material is used to tune the electronic properties and subsequent surface coverage of gold surfaces/monolayer.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ferdinand Rissner et al.
ACS nano, 4(11), 6735-6746 (2010-11-05)
The electronic structure of mixed self-assembled monolayers (SAMs) on Au(111) surfaces is modeled using slab-type density-functional theory calculations. The studied molecules have a dipolar character induced by polar and electron donating or accepting tail-group substituents. The resulting electronic structure of
Nikolaus Meyerbröker et al.
Langmuir : the ACS journal of surfaces and colloids, 28(25), 9583-9592 (2012-06-02)
Here we describe the behavior of self-assembled monolayers (SAMs) of 4'-cyanobiphenyl-4-thiol (CBPT) on Au(111) upon electron irradiation. Under such a treatment, the aromatic framework of CBPT SAMs is laterally cross-linked while the nitrile groups, located at the SAM-ambience interface, are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service