Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

757225

Sigma-Aldrich

Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4O2S2
CAS Number:
Molecular Weight:
196.25
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

form

solid

mp

274-277 °C

storage temp.

2-8°C

SMILES string

O=Cc1cc2sc(C=O)cc2s1

InChI

1S/C8H4O2S2/c9-3-5-1-7-8(11-5)2-6(4-10)12-7/h1-4H

InChI key

UBKNFAFBINVTOP-UHFFFAOYSA-N

Application

This molecule has been used in the synthesis of novel metal-free organic dyes for Dye-Sensitized Solar Cells reaching conversion efficiencies of 6.23%.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niklas Keller et al.
Nanoscale, 11(48), 23338-23345 (2019-12-04)
Covalent organic frameworks (COFs), consisting of covalently connected organic building units, combine attractive features such as crystallinity, open porosity and widely tunable physical properties. For optoelectronic applications, the incorporation of heteroatoms into a 2D COF has the potential to yield
Shao-Lu Li et al.
Chemical communications (Cambridge, England), 26(26), 2792-2794 (2006-10-03)
Two novel metal-free organic dyes containing thienothiophene and thiophene segments have been synthesized. Nano-crystalline TiO2 dye-sensitized solar cells were fabricated using these dyes as light-harvesting sensitizers, and a high solar energy-to-electricity conversion efficiency of 6.23% was achieved.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service