Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

753556

Sigma-Aldrich

3-epi-25-Hydroxyvitamin D2 solution

100 μg/mL in ethanol, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44O2
CAS Number:
Molecular Weight:
412.65
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

Assay

98% (CP)

form

solution

concentration

100 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

application(s)

detection

storage temp.

−20°C

SMILES string

C[C@H](\C=C\[C@H](C)C(C)(C)O)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C3\C[C@H](O)CCC3=C

InChI

1S/C28H44O2/c1-19-10-14-24(29)18-23(19)13-12-22-8-7-17-28(6)25(15-16-26(22)28)20(2)9-11-21(3)27(4,5)30/h9,11-13,20-21,24-26,29-30H,1,7-8,10,14-18H2,2-6H3/b11-9+,22-12+,23-13-/t20-,21+,24-,25-,26+,28-/m1/s1

InChI key

KJKIIUAXZGLUND-FOZBYKFWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-epi-25-Hydroxyvitamin D2 (C-3 epimer) is a metabolite of vitamin D2. It is an epimer of 25-hydroxyvitamin D2 which differs only at the C-3 position.[1]

Application

3-epi-25-Hydroxyvitamin D2 can be used as an analytical standard for accurate data interpretation in specific biological samples.

Packaging

1 mL in ampule

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of a new vitamin D blood test on the Emirati population
Al-Dabbagh B, et al.
The Journal of Steroid Biochemistry and Molecular Biology, 180, 118-128 (2018)

Articles

Novel sample prep technique coupled with the unique selectivity of the Ascentis Express F5 column enables a fast and simplified bioanalytical method for associated vitamin D metabolites.

Using HybridSPE-Phospholipid, Ascentis Express, and Other LC-MS Workflow Components

Polar graphitic carbon enables UHPLC separation of highly polar pesticides, herbicides, and Vitamin D metabolites, advancing liquid chromatography capabilities.

Polar graphitic carbon enables UHPLC separation of highly polar pesticides, herbicides, and Vitamin D metabolites, advancing liquid chromatography capabilities.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service