Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

752959

Sigma-Aldrich

(Tributylphosphoranylidene)acetonitrile solution

1 M in toluene

Synonym(s):

CMBP solution, Tsunoda reagent, Cyanomethylene)tributylphosphorane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H28NP
Molecular Weight:
241.35
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

1 M in toluene

refractive index

n/D 1.500 (lit.)
n20/D 1.501

density

0.886 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCCCP(CCCC)(CCCC)=CC#N

InChI

1S/C14H28NP/c1-4-7-11-16(14-10-15,12-8-5-2)13-9-6-3/h14H,4-9,11-13H2,1-3H3

InChI key

OZMLUMPWPFZWTP-UHFFFAOYSA-N

General description

(Tributylphosphoranylidene)acetonitrile is a stabilized trialkylphosphorane that can act as a substitute to the diethyl azodicarboxylate and triphenylphosphine (DEAD-TPP) system for Mitsunobu reaction between nucleophiles (HA) and alcohols (ROH) to form RA. It is highly useful when the pKa of the nucleophiles are higher than 11.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Novel reactivity of stabilized methylenetributylphosphorane: a new Mitsunobu reagent.
Tsunoda T
Tetrahedron Letters, 35(28), 5081-5082 (1994)
Application of Tsunoda Reagent to the Convenient Synthesis of Drug-Like Pyrazoles.
Mosallanejad A
Tetrahedron Letters, 59(18), 1708-1710 (2018)
Izumi Sakamoto et al.
Chemical & pharmaceutical bulletin, 53(11), 1508-1509 (2005-11-08)
(Cyanomethylene)tributylphosphorane (CMBP), which promoted the alkylation of various nucleophiles (HA) with alcohols (ROH) to give RA (Mitsunobu-type reaction), was prepared in two steps starting from chloroacetonitrile.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service