Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

751596

Sigma-Aldrich

Diethyl (difluoromethyl)phosphonate

97%

Synonym(s):

1-[Difluoromethyl(ethoxy)phosphoryl]oxyethane, Diethyl P-(difluoromethyl)phosphonate, Diethyl difluoromethanephosphonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2CHP(O)(OC2H5)2
CAS Number:
Molecular Weight:
188.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.388 (lit.)
n20/D 1.388

bp

55 °C/0.5 mmHg (lit.)

density

1.199 g/mL at 25 °C (lit.)
1.206 g/mL at 25 °C

functional group

fluoro
phosphonate

storage temp.

2-8°C

SMILES string

CCOP(=O)(OCC)C(F)F

InChI

1S/C5H11F2O3P/c1-3-9-11(8,5(6)7)10-4-2/h5H,3-4H2,1-2H3

InChI key

VVFGLBKYBBUTRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service