Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

750271

Sigma-Aldrich

Potassium trans-1-propenyltrifluoroborate

97%

Synonym(s):

Potassium (E)-trifluoro(1-propenyl)borate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5BF3K
CAS Number:
Molecular Weight:
147.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

81-85 °C

SMILES string

[K+].C\C=C\[B-](F)(F)F

InChI

1S/C3H5BF3.K/c1-2-3-4(5,6)7;/h2-3H,1H3;/q-1;+1/b3-2+;

InChI key

RLDWVFWDURMTAV-SQQVDAMQSA-N

Related Categories

Application

Potassium trans-1-propenyltrifluoroborate can be used:
  • As a substrate in the cross-coupling reactions with 2-(chloromethyl)-2,1-borazaronaphthalenes to yield allyl borazaronaphthalenes using a palladium catalyst.
  • In the synthesis of anethole dithiolethione-NH2 (ADT-NH2), which in turn is used to prepare a drug-H2S delivery system.
  • To prepare methyl (E)-2,2-dimethyl-5-(propen-1-yl)-4H-benzo[d][1,3]dioxine-7-carboxylate, a key intermediate for the synthesis of the alkaloid ampullosine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

First total synthesis of ampullosine, a unique isoquinoline alkaloid isolated from Sepedonium ampullosporum, and of the related permethylampullosine
Vargas DF, et al.
Royal Society of Chemistry Advances, 9(57), 33096-33106 (2019)
Synthesis of amino-ADT provides access to hydrolytically stable amide-coupled hydrogen sulfide releasing drug targets
Hammers MD, et al.
Synlett, 27(09), 1349-1353 (2016)
Accessing 2-(Hetero) arylmethyl-,-allyl-, and-propargyl-2, 1-borazaronaphthalenes: Palladium-Catalyzed Cross-Couplings of 2-(Chloromethyl)-2, 1-borazaronaphthalenes
Molander GA, et al.
Organic Letters, 16(22), 6024-6027 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service