Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

749427

Sigma-Aldrich

Lithium carbonate-13C

99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li213CO3
Molecular Weight:
74.88
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

crystals

mp

618 °C (lit.)

mass shift

M+1

SMILES string

[Li+].[Li+].[O-][13C]([O-])=O

InChI

1S/CH2O3.2Li/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2/i1+1;;

InChI key

XGZVUEUWXADBQD-GOCMCNPZSA-L

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service