Skip to Content
MilliporeSigma
All Photos(1)

Documents

749397

Sigma-Aldrich

Sodium D-3-hydroxybutyrate-2,4-13C2

endotoxin tested, ≥99 atom % 13C

Synonym(s):

(R)-(-)-3-Hydroxybutyric acid-2,4-13C2 sodium salt, Sodium (R)-3-hydroxybutyrate-2.4.13C2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C2H7O3Na
Molecular Weight:
128.07
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

149-155 °C (lit.)

suitability

endotoxin tested

mass shift

M+2

storage temp.

2-8°C

SMILES string

[13CH3][C@@H](O)[13CH2]C([O-])=O.[Na+]

InChI

1S/C4H8O3.Na/c1-3(5)2-4(6)7;/h3,5H,2H2,1H3,(H,6,7);/q;+1/p-1/t3-;/m1./s1/i1+1,2+1;

InChI key

NBPUSGBJDWCHKC-WGJKDMNVSA-M

Looking for similar products? Visit Product Comparison Guide

Application

For R&D use only. Not for drug, household, or other uses. Any other use is the sole responsibility of the end user.

Endotoxin testing is done on the bulk form of the product before subdivision packaging. This bulk test does not guarantee that the subdivision or repackaged aliquot is endotoxin-free when it is received or used by the customer and does not imply suitability for any particular purpose. If the product must be endotoxin-free for the intended application, the product should be tested prior to actual use.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service