Skip to Content
MilliporeSigma
All Photos(1)

Documents

749230

Sigma-Aldrich

Dichlorotetrakis[3,5-difluoro-2-(2-pyridinyl)phenyl]diiridium(III)

95%

Synonym(s):

Di-μ-chlorotetrakis[3,5-difluoro-2-(2-pyridinyl-κN)phenyl- κC]di-iridium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H24Cl2F8Ir2N4
CAS Number:
Molecular Weight:
1216.02
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid

mp

>400 °C (lit.)

SMILES string

Cl[Ir].Cl[Ir].Fc1ccc(c(F)c1)-c2ccccn2.Fc3ccc(c(F)c3)-c4ccccn4.Fc5ccc(c(F)c5)-c6ccccn6.Fc7ccc(c(F)c7)-c8ccccn8

InChI

1S/4C11H7F2N.2ClH.2Ir/c4*12-8-4-5-9(10(13)7-8)11-3-1-2-6-14-11;;;;/h4*1-7H;2*1H;;/q;;;;;;2*+1/p-2

InChI key

RXDKZUKXPWIFAN-UHFFFAOYSA-L

Application

This material is a precursor to a wide range of OLED emitters such as FIrpic.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service